• USA Home
  • 816106 - Holder for 21.5 mm I.D. TSKgel® Guard Cartridges

EMAIL THIS PAGE TO A FRIEND
816106 Supelco

Holder for 21.5 mm I.D. TSKgel® Guard Cartridges

pkg of 1 ea

Synonym: TSKgel® Holder

  •  eCl@ss 32110501

Purchase

Properties

Related Categories Analytical/Chromatography, HPLC, HPLC & FPLC Columns for Biopolymer Separations, Holders, TSKgel HPLC Columns, TOYOPEARL Bulk Media & Accessories,
packaging   pkg of 1 ea
mfr. no.   Tosoh, 16106
application(s)   HPLC: suitable

Description

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?