• USA Home
  • 817358 - TSKgel® G6000HHR HPLC Column

EMAIL THIS PAGE TO A FRIEND
817358 Supelco

TSKgel® G6000HHR HPLC Column

phase polystyrene-divinylbenzene, L × I.D. 30 cm × 7.8 mm, 5 μm particle size

Synonym: TSKgel® HHR Column

Purchase

Properties

Related Categories Analytical/Chromatography, HPLC, HPLC & FPLC Columns for Biopolymer Separations, TSKgel Gel Permeation Columns for Organic-Soluble Polymers, TSKgel HHR Type Gel Permeation Columns,
material   stainless steel hardware
mfr. no.   Tosoh, 17358
parameter   140 °C temperature
  2 mL/min flow rate
  50 bar max. pressure
L × I.D.   30 cm × 7.8 mm
matrix   polymer particle platform
matrix active group   polystyrene-divinylbenzene phase
particle size   5 μm
suitability   suitable for L21 per USP
mode of chromatography   Size exclusion (SEC)

Description

Compatibility

THF can be replaced with acetone, ethanol, quinoline, benzene, o-dichlorobenzene, ethyl acetate, dodecane, FC-113, carbon tetrachloride, dichloromethane, dichloroethane, trichloroethane, n-hexane, cyclohexane, xylene, tetrahydrofuran, chloroform, 1,4-dioxane, hexafluoroisopropanol, toluene, 1-chloronaphthalene, dimethylaceto acetamide, methyl ethyl ketone, trichlorobenzene, m-cresol, dimethylformamide, methylpyrrolidone, o-chlorophenol/chloroform, dimethyl sulfoxide, pyridine

General description

The TSKgel HHR column line consists of eight columns with different pore sizes, TSKgel G1000HHR through TSKgel G7000HHR, and five columns with an extended linear range of the calibration curve. All TSKgel HHR-type columns are shipped in THF, which can be exchanged for a much broader range of organic solvents than TSKgel HXL columns, although Hxl columns are more inert. The maximum operating temperature for most TSKgel HHR-type columns is 140°C, except for GMHHR columns, which can only be used up to 80°C. TSKgel HHR-type columns are conventional GPC columns with dimensions of 7.8mm ID x 30cm.

TSKgel G6000Hhr columns are packed with rigid, porous, polymer beads. They are used for the non-aqueous separation of polymers less than 40,000,000 Da. TSKgel G6000Hhr columns are shipped in THF, which can be exchanged for a wide variety of organic solvents.

Physical form

Shipped in THF.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

analytical column

Product #

Description

Add to Cart

817352 TSKgel® G1000HHR HPLC Column, phase polystyrene-divinylbenzene, L × I.D. 30 cm × 7.8 mm, 5 μm particle size
817354 TSKgel® G2500HHR HPLC Column, phase polystyrene-divinylbenzene, L × I.D. 30 cm × 7.8 mm, 5 μm particle size
817357 TSKgel® G5000HHR HPLC Column, phase polystyrene-divinylbenzene, L × I.D. 30 cm × 7.8 mm, 5 μm particle size
817392 TSKgel® GMHHR-M HPLC Column, phase polystyrene-divinylbenzene, L × I.D. 30 cm × 7.8 mm, 5 μm particle size
818055 TSKgel® GMHHR-N HPLC Column, phase polystyrene-divinylbenzene, L × I.D. 30 cm × 7.8 mm, 5 μm particle size

guard column

Product #

Description

Add to Cart

817369 TSKgel® HHR Guard Column, phase polystyrene-divinylbenzene, L × I.D. 4 cm × 6 mm, 5 μm particle size

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?