• USA Home
  • 817369 - TSKgel® HHR Guard Column

EMAIL THIS PAGE TO A FRIEND
817369 Supelco

TSKgel® HHR Guard Column

phase polystyrene-divinylbenzene, L × I.D. 4 cm × 6 mm, 5 μm particle size

Synonym: TSKgel® HHR Column

Purchase

Properties

Related Categories Analytical/Chromatography, HPLC, HPLC & FPLC Columns for Biopolymer Separations, TSKgel Gel Permeation Columns for Organic-Soluble Polymers, TSKgel HHR Type Gel Permeation Columns,
mfr. no.   Tosoh, 17369
parameter   0.5-1.0 mL/min standard flow rate
  80 °C temperature
L × I.D.   4 cm × 6 mm
matrix   polymer particle platform
matrix active group   polystyrene-divinylbenzene phase
particle size   5 μm
suitability   suitable for L21 per USP
mode of chromatography   Size exclusion (SEC)

Description

Compatibility

THF can be replaced with acetone, ethanol, quinoline, benzene, o-dichlorobenzene, ethyl acetate, dodecane, FC-113, carbon tetrachloride, dichloromethane, dichloroethane, trichloroethane, n-hexane, cyclohexane, xylene, tetrahydrofuran, chloroform, 1,4-dioxane, hexafluoroisopropanol, toluene, 1-chloronaphthalene, dimethylaceto acetamide, methyl ethyl ketone, trichlorobenzene, m-cresol, dimethylformamide, methylpyrrolidone, o-chlorophenol/chloroform, dimethyl sulfoxide, pyridine

General description

Guard Column for TSKgel G5000Hhr-G7000Hhr & GMHhr-M;-N;-H columns, 5μm

The TSKgel HHR column line consists of eight columns with different pore sizes, TSKgel G1000HHR through TSKgel G7000HHR, and five columns with an extended linear range of the calibration curve. All TSKgel HHR-type columns are shipped in THF, which can be exchanged for a much broader range of organic solvents than TSKgel HXL columns, although Hxl columns are more inert. The maximum operating temperature for most TSKgel HHR-type columns is 140°C, except for GMHHR columns, which can only be used up to 80°C. TSKgel HHR-type columns are conventional GPC columns with dimensions of 7.8mm ID x 30cm.

Physical form

Shipped in THF.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?