861238 Supelco

EPA 8270 Internal Standard Mix 2

2000 μg/mL each component in methylene chloride: benzene (1:1), analytical standard

DOWNLOAD MSDS (PDF)

Properties

Related Categories 8000 Series Solidwaste Methods, Analytical Standards, Analytical/Chromatography, EPA, Method 8270,
grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
feature   standard type internal
packaging   ampule of 100 mL
concentration   2000 μg/mL each component in methylene chloride: benzene (1:1)
suitability   suitable for 8270 per US EPA
  suitable for OLM04 Statement of Work per US EPA

Price and Availability

Analyte

Description

Product #

Add to Cart

Acenaphthene-d10    
Chrysene-d12    
1,4-Dichlorobenzene-d4    
Naphthalene-d8    
Perylene-d12    
Phenanthrene-d10    

Customers Also Viewed

1,4-<WBR>Dichlorobenzene-<WBR>d<SUB>4</SUB>

98% D, 98% (CP)

(each component at wt% indicated), analytical standard

500 μg/mL each component in acetone, analytical standard

2000 μg/mL each component in methylene chloride, analytical standard

4000 μg/mL each component in methylene chloride: benzene (1:1), analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?