861252 Supelco

EPA 8270 Base Neutral Surrogate Mix 1

2000 μg/mL each component in methylene chloride: benzene (1:1), analytical standard

DOWNLOAD MSDS (PDF)

Properties

Related Categories 8000 Series Solidwaste Methods, Analytical Standards, Analytical/Chromatography, EPA, Method 8270,
grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
feature   standard type surrogate
packaging   ampule of 100 mL
concentration   2000 μg/mL each component in methylene chloride: benzene (1:1)
suitability   suitable for 8270 per US EPA

Price and Availability

Analyte

Description

Product #

Add to Cart

2-Fluorobiphenyl    
Nitrobenzene-d5    
p-Terphenyl-d14    

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?