• USA Home
  • 8S61148 - EPA 8270 Calibration Mix 4

EMAIL THIS PAGE TO A FRIEND
8S61148 Supelco

EPA 8270 Calibration Mix 4

certified reference material, 2000 μg/mL each component in methylene chloride

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Certified Reference Materials (CRMs), Chromatography,
grade   certified reference material
description   Separate Source
CofA   certificate of analysis is enclosed in each package.
packaging   ampule of 1 × 1 mL
concentration   2000 μg/mL each component in methylene chloride
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
suitability   suitable for 8270 per US EPA
format   multi-component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Analyte

Description

Product #

Add to Cart

Aniline SDS    
Benzoic acid SDS    
Benzyl alcohol SDS    
Pyridine SDS    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1593 6.1 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?