EMAIL THIS PAGE TO A FRIEND
8S61323 Supelco

EPA 8260 Mix 5

analytical standard, in methanol: water (9:1) (varied)

Purchase

Properties

Related Categories 8000 Series Solidwaste Methods, Additional Standards, Analytical Standards, Analytical/Chromatography, Chromatography,
grade   analytical standard
description   Separate Source
CofA   certificate of analysis is enclosed in each package.
feature   standard type calibration
packaging   ampule of 1 × 1 mL
concentration   in methanol: water (9:1) (varied)
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
suitability   suitable for 8260 per US EPA
format   multi-component solution
storage temp.   −20°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Analyte

Description

Product #

Add to Cart

Acetone 2000 μg/mL SDS    
tert-Butanol 40000 μg/mL SDS    
2-Butanone 2000 μg/mL SDS    
tert-Butyl methyl ether 2000 μg/mL SDS    
Carbon disulfide 2000 μg/mL SDS    
2-Hexanone 2000 μg/mL SDS    
4-Methyl-2-pentanone 2000 μg/mL SDS    
Vinyl acetate 2000 μg/mL SDS    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?