• USA Home
  • 97718-U - SupelFlash Cartridge
EMAIL THIS PAGE TO A FRIEND
97718-U Supelco
SupelFlash Cartridge
matrix active group silica, tube volume 150 mL, bed wt. 70 g, 40-63 μm, pkg of 8
Purchase
Properties
composition   bed wt., 70 g
packaging   pkg of 8
tube volume   150 mL
matrix active group   silica
particle size   40-63 μm
pore size   60 Å
compatibility   for use with Biotage Flashmaster systems
Description
General description
SupelFlash Cartridges
SupelFlash cartridges are pre-compressed, disposable flash columns containing ultrapure silica, designed to meet your stringent flash purification requirements. Cartridge pre-compression reduces particle interstitial spacing to provide even mass transfer and balanced chemical interaction.
Features Include:
Female luer lock inlet and male slip luer outlet for easy compatibility
8 Bed weights from 4 grams to 330 grams
40-60 um irregular silica
Higher efficiency leading to sharper more symmetric peaks
High yield and sample loading capacity
Reliable, consistent purification performance
Supel Flash Cartridges are Designed For Use in:
ISCO’s CombiFlash, Companion, Retrieve, and OptiX systems;
Analogix’s SimpliFlash, and IntelliFlash and Biotage systems.
Legal Information
FlashMaster is a trademark of Biotage AB
Supel is a trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?