• USA Home
  • 97766-U - Solid Sample Cartridge Replacement Frits

EMAIL THIS PAGE TO A FRIEND
97766-U Supelco

Solid Sample Cartridge Replacement Frits

size 40 mm

Synonym: VersaFlash® Solid Sample Cartridge Frit

Purchase

Description

Legal Information

VersaFlash is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability


All labs need water

Non-Sterile Millex Syringe Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?