• USA Home
  • 97783-U - VersaPak® Silica Cartridge
EMAIL THIS PAGE TO A FRIEND
97783-U Supelco
VersaPak® Silica Cartridge
20-45 μm, matrix active group Spherical Silica, bed wt. 20 g, tube volume 60 mL, pkg of 10
Purchase
Properties
composition   bed wt., 20 g
packaging   pkg of 10
tube volume   60 mL
matrix active group   Spherical Silica
particle size   20-45 μm
compatibility   for use with Biotage FlashMaster
systems
Description
Legal Information
FlashMaster is a trademark of Biotage AB
VersaPak is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
All labs need water
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?