Supelco

EPA 524.2 Rev 4 Update Ketones Mix

analytical standard, 2000 μg/mL in methanol: water (9:1)

Properties

Related Categories 500 Series Drinking Water Methods, Analytical Standards, Analytical/Chromatography, Chromatography, Drinking Water,
grade   analytical standard
description   Separate Source
CofA   certificate of analysis is enclosed in each package.
feature   standard type calibration
packaging   ampule of 1 × 1 kit (4S7428-U)
  ampule of 1 × 1 mL (47428-U)
concentration   2000 μg/mL in methanol: water (9:1)
suitability   suitable for 524.2 per US EPA
storage temp.   2-8°C

Price and Availability

Analyte

Description

Product #

Add to Cart

Acetone    
2-Butanone    
1,1-Dichloro-2-propanone    
2-Hexanone    
4-Methyl-2-pentanone    

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?