Supelco

EPA VOC Mix 5

analytical standard, 2000 μg/mL each component in methanol

  •  EC Number 200-659-6

Properties

Related Categories 500 Series Drinking Water Methods, 8000 Series Solidwaste Methods, Alpha Sort, Alphabetical Index of Analytical Standards, Analytical Standards,
grade   analytical standard
description   Separate Source
CofA   certificate of analysis is enclosed in each package.
feature   standard type calibration
packaging   ampule of 1 × 1 mL (48797)
  ampule of 1 × 1 mL (4S8797)
concentration   2000 μg/mL each component in methanol
suitability   suitable for 502.1 per US EPA
  suitable for 502.2 per US EPA
  suitable for 524.1 per US EPA
  suitable for 524.2 per US EPA
  suitable for 8010 per US EPA
  suitable for 8021 (1987) per US EPA
  suitable for 8021A (1990) per US EPA
  suitable for 8260 per US EPA
  suitable for Group 16 per MISA (Canada)
storage temp.   2-8°C

Price and Availability

Analyte

Description

Product #

Add to Cart

Bromodichloromethane    
Dibromochloromethane    
1,1-Dichloroethylene    
cis-1,2-Dichloroethylene    
trans-1,2-Dichloroethylene    
Dichloromethane    

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?