Supelco

EPA TCL Pesticides Mix

analytical standard, 2000 μg/mL each component in hexane: toluene (1:1)

Properties

Related Categories 8000 Series Solidwaste Methods, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Canada,
grade   analytical standard
description   Separate Source
CofA   certificate of analysis is enclosed in each package.
feature   standard type calibration
packaging   ampule of 1 × 1 mL (48913)
  ampule of 1 × 1 mL (4S8913)
concentration   2000 μg/mL each component in hexane: toluene (1:1)
suitability   suitable for 8081 per US EPA
  suitable for 8270 per US EPA
  suitable for Group 22 per MISA (Canada)
storage temp.   2-8°C

Price and Availability

Analyte

Description

Product #

Add to Cart

Aldrin    
α-BHC    
β-BHC    
Lindane    
δ-BHC    
1,1-Dichloro-2,2-bis(4-chlorophenyl)ethane    
4,4′-DDE    
4,4′-DDT    
Dieldrin    
α-Endosulfan    
β-Endosulfan    
Endosulfan sulfate    
Endrin    
Endrin aldehyde    
Endrin ketone    
Heptachlor    
Heptachlor exo-epoxide    
Methoxychlor    

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?