| Related Categories | All Pharmaceutical Applications, Analytical/Chromatography, Antihypertensives, Applications, Chiral Applications, |
| test parameters | column: CHIROBIOTIC T2, 25 cm x 4.6 mm I.D., 5 μm particles (16024AST) |
| mobile phase: 5 mM ammonium formate in methanol | |
| flow rate: 1 mL/min | |
| column temp.: 25 °C | |
| detector: UV, 220 nm | |
| injection: 5 μL | |
| sample: 1 mg/mL in methanol |
Astec is a registered trademark of Sigma-Aldrich Co. LLC
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC
|
Description |
Product # |
Add to Cart |
|---|---|---|
| Oxprenolol hydrochloride |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?