CAS Number 60-51-5
Empirical Formula (Hill Notation) C5H12NO3PS2
Molecular Weight 229.26
Beilstein Registry Number 1785339
EC Number 200-480-3
MDL number MFCD00053676
PubChem Substance ID 24899264
| grade | analytical standard |
| packaging | ampule of 250 mg |
| mfr. no. | Chem Service Inc, PS-659 |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?