| Related Categories | Analytical/Chromatography, Sampling, Storage, and Serum Bottles, Serum Bottles, 5 mL to 100 mL, Supelco glass serum bottles, Vials More... |
| material | clear glass |
| feature | autoclavable |
| closure type 20mm crimp seal (not included) | |
| packaging | pkg of 1 × 144 ea (33111-U) |
| pkg of 1 × 36 ea (33110-U) | |
| O.D. × H | 51.7 mm × 94.5 mm |
| size | 100 mL |
These vials are constructed of clear, borosilicate glass; closure not included.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?