Supelco

EPA 501/601 Trihalomethanes Calibration Mix

2000 μg/mL each component in methanol, analytical standard

Properties

Related Categories 500 Series Drinking Water Methods, 600 Series Wastewater Methods, Analytical Standards, Analytical/Chromatography, EPA,
grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
feature   standard type calibration
packaging   ampule of 1 × 1 mL (48140-U)
  ampule of 5 × 1 mL (4M8140-U)
concentration   2000 μg/mL each component in methanol
suitability   suitable for 501.1 per US EPA
  suitable for 501.2 per US EPA
  suitable for 501.3 per US EPA
  suitable for 601 per US EPA
storage temp.   2-8°C

Price and Availability

Analyte

Description

Product #

Add to Cart

Bromodichloromethane    
Bromoform    
Chloroform    
Dibromochloromethane    

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?