• USA Home
  • ES5679 - ChromegaChiral Chiral CC3, 3 Micron HPLC Column
EMAIL THIS PAGE TO A FRIEND
ES5679 Supelco
ChromegaChiral Chiral CC3, 3 Micron HPLC Column
3 μm particle size, L × I.D. 25 cm × 4.6 mm
Purchase
Properties
Related Categories Analytical/Chromatography, Chiral HPLC Columns, ChromegaChiral CC3 3 Micron Chiral Columns, ChromegaChiral CC3 Chiral Columns, ChromegaChiral Columns,
mfr. no.   ES Industries, 155151-CC3
L × I.D.   25 cm × 4.6 mm
matrix active group   amylose, carbamate phase
particle size   3 μm
pore size   1000 Å pore size
mode of chromatography   Chiral
Description
General description
ChromegaChiral CC3 (amylose tris(5-chloro-2-methyl- phenylcarbamate) is a new product for high resolution chiral separations based on a new halogenated carbohy- drate based chiral stationary phase.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
ChromegaChiral is a trademark of ES Industries
Price and Availability
All labs need water
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Content
Supelco HPLC Calculator for Isocratic or Gradient Method Transfer
Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?