• USA Home
  • FFA57287-U - SPME fiber assortment kit 4

EMAIL THIS PAGE TO A FRIEND
FFA57287-U Supelco

SPME fiber assortment kit 4

for use with multi fiber exchanger, needle size 23 ga

Purchase

Properties

Related Categories Analytical/Chromatography, SPME FFA Assortment Kits, SPME Fast Fit Fiber Assemblies (FFA) & Accessories, SPME Fibers and Holders, Sample Preparation & Purification,
material   fused silica fiber
needle size   23 ga
df   100 μm (PDMS)
  65 μm (PDMS/DVB)
  75 μm (CAR/PDMS)
matrix active group   Carboxen/Polydimethylsiloxane (CAR/PDMS) coating, 75 μm
  Polydimethylsiloxane (PDMS) coating, 100 μm
  Polydimethylsiloxane/Divinylbenzene (PDMS/DVB) coating, 65 μm
compatibility   for analyte group flavors and odors
  for use with multi fiber exchanger

Description

Components

Kit 4 - For Flavors and Odors - contains one of each:
• 100μm polydimethylsiloxane coating
• 65μm polydimethylsiloxane/divinylbenzene coating
• 75μm Carboxen/polydimethylsiloxane coating

Price and Availability


All labs need water

Non-Sterile Millex Syringe Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Fundamentos basicos de microextraccion en fase solida (SPME) y sus aplicaciones [Webinar]

En este webinar presentamos los fundamentos básicos de microextracción en fase sólida (SPME) y sus aplicaciones. SPME es una técnica para análisis de compuestos volátiles y semi-volátiles, fácil de u...
Keywords: Solid phase microextractions

SUPELCO® SPME Fast-Fit Fiber Assemblies [VIDEO]

Ideal for SPME method development, SPME Fast-Fit Fiber Assemblies (FFA) from Supelco are  easy to install, feature a non-threaded attachment, and are equipped with a traceable color-coded barcode lab...
Keywords: Solid phase microextractions

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?