• USA Home
  • G002444 - LC/MS Analysis of Peptides, Basic, on Ascentis® C18

EMAIL THIS PAGE TO A FRIEND
G002444 Supelco

LC/MS Analysis of Peptides, Basic, on Ascentis® C18

application for HPLC

Properties

Related Categories Analytical/Chromatography, Applications, HPLC Applications, Peptides
test parameters   column: Ascentis C18, 10 cm x 2.1 mm I.D., 5 μm particles (581326-U)
  mobile phase: [A] 20 mM formic acid in water; [B]: 50:50 (v/v) 20 mM formic acid in water:20 mM formic acid in acetonitrile
  gradient: 20 to 60% B in 20 min
  flow rate: 0.2 mL/min
  column temp.: 22 °C
  detector: ESI (+), full scan
  injection: 2 μL
suitability   application for HPLC
Featured Industry   Life Science and Biopharma

Price and Availability

Description

Legal Information

Ascentis is a registered trademark of Sigma-Aldrich Co. LLC

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?