• USA Home
  • G002597 - HPLC Analysis of Catechols and Resorcinols on Ascentis® RP-Amide

EMAIL THIS PAGE TO A FRIEND
G002597 Supelco

HPLC Analysis of Catechols and Resorcinols on Ascentis® RP-Amide

application for HPLC

Properties

Related Categories Analytical/Chromatography, Applications, Clinical, HPLC Applications, Phenols,
test parameters   column: Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 μm particles (565324-U)
  mobile phase: [A] 20 mM phosphoric acid (pH 2.0 unadjusted): [B] acetonitrile; (75:25, A:B)
  flow rate: 1.5 mL/min
  column temp.: 30 °C
  detector: UV, 270 nm
  injection: 25 μL
  sample: as indicated in 20 mM phosphoric acid (pH 2.0)
suitability   application for HPLC
Featured Industry   Chemicals and Industrial Polymers
Clinical

Price and Availability

Description

Legal Information

Ascentis is a registered trademark of Sigma-Aldrich Co. LLC

Analyte

Description

Product #

Add to Cart

Resorcinol 50 mg/mL SDS    
Pyrocatechol SDS    
2-Methylresorcinol SDS    
4-Methylcatechol SDS    
2,5-Dimethylresorcinol 50 mg/mL SDS    
3-Methylcatechol 50 mg/mL SDS    
4-Nitrocatechol 50 mg/mL SDS    

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?