• USA Home
  • G002688 - LC/MS Analysis of Peptides, Hydrophobic, on Ascentis® C18

EMAIL THIS PAGE TO A FRIEND
G002688 Supelco

LC/MS Analysis of Peptides, Hydrophobic, on Ascentis® C18

application for HPLC

Properties

Related Categories Analytical/Chromatography, Applications, HPLC Applications, Peptides
test parameters   column: Ascentis C18, 10 cm x 2.1 mm I.D., 5 μm particles (581326-U)
  mobile phase: [A] 50:50 (v/v) 50 mM formic acid, pH 3.0 with acetic acid:water; [B] 50:50 (v/v) 50 mM formic acid, pH 3.0 with acetic acid:acetonitrile
  gradient: 20 to 50% B in 15 min
  flow rate: 0.2 mL/min
  column temp.: ambient
  detector: ESI (+)
  injection: 4 μL
  sample: as indicated in 10 mM formic acid
suitability   application for HPLC
Featured Industry   Life Science and Biopharma

Price and Availability

Description

Legal Information

Ascentis is a registered trademark of Sigma-Aldrich Co. LLC

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?