• USA Home
  • G003035 - HPLC Analysis of Vitamins, Water Soluble, on Ascentis® RP-Amide

EMAIL THIS PAGE TO A FRIEND
G003035 Supelco

HPLC Analysis of Vitamins, Water Soluble, on Ascentis® RP-Amide

application for HPLC

Properties

Related Categories Analytical/Chromatography, Anticonvulsant/antiepiletic/antiseizure, Applications, Biochemicals and Reagents, Chromatographic Methods for Water Soluble Vitamin Analysis,
test parameters   column: Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 μm particles (565324-U)
  mobile phase: 25 mM potassium phosphate, dibasic (pH 3.5 with phosphoric acid)
  flow rate: 1.0 mL/min
  column temp.: 30 °C
  detector: UV, 230 nm
  injection: 10 μL
  sample: as indicated in 25 mM potassium phosphate, dibasic (pH 3.5 with phosphoric acid)
suitability   application for HPLC
Featured Industry   Vitamins, Nutraceuticals, and Natural Products

Price and Availability


All labs need water

Description

Legal Information

Ascentis is a registered trademark of Sigma-Aldrich Co. LLC

Analyte

Description

Product #

Add to Cart

Niacin 50 μg/mL SDS    
L-Ascorbic acid SDS    
Pyridoxine SDS    
Thiamine 75 μg/mL SDS    

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?