• USA Home
  • G003531 - US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SLB®-5ms

EMAIL THIS PAGE TO A FRIEND
G003531 Supelco

US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SLB®-5ms

application for GC

Properties

Related Categories Analytical/Chromatography, Applications, Capillary GC Columns and Guard Columns/Retention Gaps, Capillary GC Columns, by industry / application, Capillary GC Columns, by method reference,
test parameters   column: SLB-5ms, 30 m x 0.25 mm I.D., 0.25 μm (28471-U)
  oven: 100 °C (2 min), 15 °C/min to 300 °C (5 min)
  inj. temp.: 250 °C
  detector: ECD, 300 °C
  carrier gas: helium, 0.9 mL/min, constant flow
  injection: 2.0 μL, splitless (0.75 min)
  liner: 4 mm I.D., single taper
  sample: chlorinated pesticide standard (46845-U), diluted to 50 ppb in n-hexane
suitability   application for GC
Featured Industry   Environmental

Price and Availability

Description

Analyte

Description

Product #

Add to Cart

α-BHC SDS    
β-BHC SDS    
γ-BHC, certified reference material, 1000 μg/mL in methanol SDS 40102  
δ-BHC SDS    
Heptachlor SDS    
Aldrin SDS    
Heptachlor epoxide, analytical standard, isomer B SDS 49042  
γ-Chlordane SDS    
α-Chlordane SDS    
1,1-Dichloro-2,2-bis(4-chlorophenyl)ethene SDS    
Dieldrin SDS    
Endrin SDS    
4,4′-DDD, PESTANAL®, analytical standard SDS 35486  
Endrin aldehyde SDS    
4,4′-DDT SDS    
Endosulfan sulfate SDS    
Methoxychlor SDS    
Endrin ketone SDS    
Decachlorobiphenyl SDS    

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?