• USA Home
  • G003701 - HPLC Analysis of Drugs of Abuse on Ascentis® Phenyl

EMAIL THIS PAGE TO A FRIEND
G003701 Supelco

HPLC Analysis of Drugs of Abuse on Ascentis® Phenyl

application for HPLC

Properties

Related Categories Analgesics, Analytical/Chromatography, Applications, HPLC Applications
test parameters   column: Ascentis Phenyl, 15 cm x 4.6 mm I.D., 5 μm particles (581616-U)
  mobile phase: [A] water with 0.1% ammonium acetate, pH 6.6, unadjusted; [B] acetonitrile with 0.1% ammonium acetate
  gradient: 25 to 40% B in 7 min; to 25% B in 1 min
  flow rate: 1.0 mL/min
  column temp.: 35 °C
  detector: UV, 280 nm
  injection: 10 μL
  sample: as indicated in mobile phase A
suitability   application for HPLC
Featured Industry   Forensics and Toxicology

Price and Availability

Description

Legal Information

Ascentis is a registered trademark of Sigma-Aldrich Co. LLC

Analyte

Description

Product #

Add to Cart

Morphine SDS    
Hydromorphone 50 mg/mL SDS    
Codeine 50 mg/mL SDS    
Hydrocodone SDS    

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?