• USA Home
  • G003712 - HPLC Analysis of Animal Feed Additives on Ascentis® Phenyl

EMAIL THIS PAGE TO A FRIEND
G003712 Supelco

HPLC Analysis of Animal Feed Additives on Ascentis® Phenyl

application for HPLC

Properties

Related Categories Analytical/Chromatography, Applications, HPLC Applications, Preservatives
test parameters   column: Ascentis Phenyl, 15 cm x 4.6 mm I.D., 5 μm particles (581616-U)
  mobile phase: [A] 10 mM ammonium formate (pH 3.0 with formic acid); [B] methanol; (78:22, A:B)
  flow rate: 1.0 mL/min
  column temp.: 35 °C
  detector: UV, 225 nm
  injection: 5 μL
  sample: as indicated in mobile phase A
suitability   application for HPLC
Featured Industry   Agriculture

Price and Availability

Description

Legal Information

Ascentis is a registered trademark of Sigma-Aldrich Co. LLC

Analyte

Description

Product #

Add to Cart

4-Hydroxyphenolarsonic acid 140 mg/mL SDS    
Roxarsone, VETRANAL, analytical standard SDS 46724  

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?