• USA Home
  • G003719 - HPLC Analysis of Deoxyribonucleosides on Ascentis® Si

EMAIL THIS PAGE TO A FRIEND
G003719 Supelco

HPLC Analysis of Deoxyribonucleosides on Ascentis® Si

application for HPLC

Properties

Related Categories Analytical/Chromatography, Applications, HPLC Applications, Nucleic Acids
test parameters   column: Ascentis Si, 10 cm x 2.1 mm I.D., 5 μm particles (581508-U)
  mobile phase: [A] 150 mM ammonium formate (pH 3.0, with formic acid); [B] acetonitrile; (10:90, A:B)
  flow rate: 0.2 mL/min
  column temp.: 35 °C
  detector: UV, 270 nm
  injection: 1 μL
  sample: as indicated, in mobile phase
suitability   application for HPLC
Featured Industry   Life Science and Biopharma

Price and Availability

Description

Legal Information

Ascentis is a registered trademark of Sigma-Aldrich Co. LLC

Analyte

Description

Product #

Add to Cart

Thymidine .1 mg/mL SDS 89270  
2′-Deoxyadenosine monohydrate, ≥98.0% (HPLC) SDS    
2′-Deoxyguanosine monohydrate, ≥98.0% (HPLC) SDS 31070  
2′-Deoxycytidine, ≥99% (HPLC) .1 mg/mL SDS D3897  

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?