• USA Home
  • G003722 - HPLC Analysis of Amino Acids, Polar Neutral and Acidic on Ascentis® Si

EMAIL THIS PAGE TO A FRIEND
G003722 Supelco

HPLC Analysis of Amino Acids, Polar Neutral and Acidic on Ascentis® Si

application for HPLC

Properties

Related Categories Amino Acid Analysis, Amino Acids, Analytical/Chromatography, Application Notes for amino acid analysis, Applications,
test parameters   column: Ascentis Si, 10 cm x 2.1 mm I.D., 5 μm particles (581508-U)
  mobile phase: [A] 100 mM ammonium formate, pH 3.0, with formic acid; [B] water; [C] acetonitrile
  gradient: 5% A constant; 10% B, 85% C to 16% B, 79% C in 20 min
  flow rate: 0.3 mL/min
  column temp.: 35 °C
  detector: ESI (+), full scan
  injection: 2 μL
  sample: as indicated, in 10:90, (50 mM ammonium formate/formic acid, (pH 3.0):acetonitrile
suitability   application for HPLC
Featured Industry   Life Science and Biopharma

Price and Availability

Description

Legal Information

Ascentis is a registered trademark of Sigma-Aldrich Co. LLC

Analyte

Description

Product #

Add to Cart

Threonine .1 mg/mL SDS    
L-Serine SDS    
L-Asparagine SDS    
Glutamine .1 mg/mL SDS    
L-Glutamic acid SDS    
Aspartic acid .1 mg/mL SDS    

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?