• USA Home
  • G003727 - HPLC Analysis of Vitamins, Fat Soluble, on Ascentis® Si

EMAIL THIS PAGE TO A FRIEND
G003727 Supelco

HPLC Analysis of Vitamins, Fat Soluble, on Ascentis® Si

application for HPLC

Properties

Related Categories Analytical/Chromatography, Applications, Biochemicals and Reagents, Chromatographic Methods for Fat Soluble Vitamin Analysis, HPLC Applications,
test parameters   column: Ascentis Si, 15 cm x 4.6 mm I.D., 5 μm particles (581512-U)
  mobile phase: [A] hexane; [B] ethyl acetate
  gradient: 10 to 30% B in 10 min; held for 2 min; to 10% B in 1 min
  flow rate: 1.0 mL/min
  column temp.: 30 °C
  detector: UV, 290 nm
  injection: 10 μL
  sample: as indicated in 96:4 (hexane:isopropanol)
suitability   application for HPLC
Featured Industry   Vitamins, Nutraceuticals, and Natural Products

Price and Availability


All labs need water

Description

Legal Information

Ascentis is a registered trademark of Sigma-Aldrich Co. LLC

Analyte

Description

Product #

Add to Cart

(±)-α-Tocopherol, analytical standard 100 mg/mL SDS 47783  
Menadione (K3), analytical standard SDS 47775  
(+)-γ-Tocopherol, analytical standard 200 mg/mL SDS 47785  
Cholecalciferol (D3), analytical standard SDS 47763  

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?