• USA Home
  • G006241 - GC Analysis of Xylene Isomers on SLB®-IL60

EMAIL THIS PAGE TO A FRIEND
G006241 Supelco

GC Analysis of Xylene Isomers on SLB®-IL60

application for GC

Properties

Related Categories Analytical/Chromatography, Applications, Aromatics, Capillary GC Columns and Guard Columns/Retention Gaps, Capillary GC Columns, by industry / application,
test parameters   column: SLB-IL60, 30 m x 0.25 mm I.D., 0.20 μm (29505-U)
  oven: 40 °C (4 min), 8 °C/min to 200 °C (5 min)
  inj. temp.: 250 °C
  detector: FID, 250 °C
  carrier gas: 30 cm/sec constant pressure
  injection: 1 μL100:1 split
  liner: 4 mm I.D., split/splitless type, wool packed single taper FocusLiner design
  sample: 12 analytes, each at 0.2% (v/v) in pentane
suitability   application for GC
Featured Industry   Chemicals and Industrial Polymers

Price and Availability

Description

Analysis Note

The xylene isomers are precursors to many chemicals:
• o-xylene is a precursor for phthalic anhydride
• m-xylene is a precursor for isophthalic acid
• p-xylene is a precursor for terephthalic acid and dimethyl terephthalate

The cresol (methylphenol) isomers are also precursors to many chemicals. This chromatogram of a mix of aromatic and methylphenol compounds was generated using an SLB-IL60 ionic liquid column. Its interaction mechanisms allow the separation of all three xylene isomers, and all three cresol isomers.

Legal Information

FocusLiner is a trademark of SGE Analytical Science Pty Ltd.

SLB is a registered trademark of Sigma-Aldrich Co. LLC

Analyte

Description

Product #

Add to Cart

Benzene SDS    
Toluene SDS    
Ethylbenzene SDS    
p-Xylene SDS    
m-Xylene SDS    
Chlorobenzene SDS    
o-Xylene SDS    
Styrene SDS    
1,4-Dichlorobenzene SDS    
o-Cresol SDS    
m-Cresol SDS    
p-Cresol SDS    

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?