• USA Home
  • LOE17AB - LUBRICATING OIL-Ba,Ca,Cl,P,S,Zn

EMAIL THIS PAGE TO A FRIEND
LOE17AB Supelco

LUBRICATING OIL-Ba,Ca,Cl,P,S,Zn

analytical standard

Synonym: Lubricating Oil w/Elements Standard, Lubricating oil with metals standard

Purchase

Properties

Related Categories ASTM D4927, ASTM D6443, ASTM D6481, ASTM Petroleum Standards, Analytical Standards,
grade   analytical standard
packaging   set of 17 × 100 mL
suitability   suitable for D4927 per ASTM
  suitable for D6443 per ASTM
  suitable for D6481 per ASTM
storage temp.   room temp

Description

General description

A number of petroleum based oil products are formulated with organo-metallic additives. These additives function as detergents, antioxidants and/or anti-wear agents. Supelco and Analytical Services (TX) have partnered to provide XRF calibration standards to measure the amount of these additives in newly manufactured oil products, additives and additive packages. Each standard is accompanied with a detailed certificate of analysis (COA).

Same as LOE17, but also contains Barium and Chlorine. 17 standards per set, 100 mL each.

Price and Availability


Nylon Membrane and Net Filters
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?