• USA Home
  • ME7 - Fatty acid methyl esters, saturated

EMAIL THIS PAGE TO A FRIEND
ME7 Supelco

Fatty acid methyl esters, saturated

~99% (unless otherwise indicated, TLC and/or GLC)

Purchase

Properties

assay   ~99% (unless otherwise indicated, TLC and/or GLC)
storage temp.   2-8°C

Description

Packaging

::FM7 High molecular weight fatty acid methyl esters, 100 mg of each

P 9036   Pentacosanoic Acid Methyl Ester
H 6389   Hexacosanoic Acid Methyl Ester
H 6639   Heptacosanoic Acid Methyl Ester
O 4129   Octacosanoic Acid Methyl Ester
N 2762   Nonacosanoic Acid Methyl Ester
T 1902   Triacontanoic Acid Methyl Ester
H 6014   Hentriacontanoic Acid Methyl Ester,
                    Approx. 98%

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?