EMAIL THIS PAGE TO A FRIEND
MET1075A Supelco

1,2,4-Triazole

analytical standard

  • CAS Number 288-88-0

  • Empirical Formula (Hill Notation) C2H3N3

  • Molecular Weight 69.07

  •  Beilstein Registry Number 104767

  •  EC Number 206-022-9

  •  MDL number MFCD00005228

  •  PubChem Substance ID 24897377

Purchase

Properties

Related Categories All Herbicide Standards, Alpha Index of All Pesticide Standards, Amitrol metabolite, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
InChI Key   NSPMIYGKQJPBQR-UHFFFAOYSA-N
form   neat
packaging   ampule of 1000 mg
mfr. no.   Chem Service, Inc., MET-1075A
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   260 °C(lit.)
mp   119-121 °C(lit.)
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
2
RTECS 
XZ3806000
Flash Point(F) 
338 °F
Flash Point(C) 
170 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?