• USA Home
  • MET543B - Methiocarb sulfoxide

EMAIL THIS PAGE TO A FRIEND
MET543B Supelco

Methiocarb sulfoxide

100 μg/mL in toluene, analytical standard

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Carbamates, Insecticides, Methiocarb metabolites,
grade   analytical standard
packaging   ampule of 5 mL
mfr. no.   Chem Service, Inc., MET-543B
concentration   100 μg/mL in toluene
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1/PG 2
WGK Germany 
3
RTECS 
FC5075000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?