• USA Home
  • MET607A - 4-tert-Butyl-2-chlorophenol

EMAIL THIS PAGE TO A FRIEND
MET607A Supelco

4-tert-Butyl-2-chlorophenol

analytical standard

  • CAS Number 98-28-2

  • Empirical Formula (Hill Notation) C10H13ClO

  • Molecular Weight 184.66

  •  EC Number 202-652-3

Purchase

Properties

Related Categories A-B, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, Pesticide Metabolite Standards,
grade   analytical standard
form   neat
packaging   ampule of 10 mg
mfr. no.   Chem Service, Inc., MET-607A
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?