• USA Home
  • MET754B - 2,3-Dihydro-2,2-dimethyl-benzofuran-7-ol

EMAIL THIS PAGE TO A FRIEND
MET754B Supelco

2,3-Dihydro-2,2-dimethyl-benzofuran-7-ol

analytical standard

Purchase

Properties

Related Categories 2,4-D metabolites, A-D, All Herbicide Standards, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
form   neat
packaging   ampule of 1000 mg
mfr. no.   Chem Service, Inc., MET-754B
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
3
RTECS 
DF7708985
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?