EMAIL THIS PAGE TO A FRIEND
MET754E Supelco

Carbofuran-3-keto

analytical standard

  • CAS Number 16709-30-1

  • Empirical Formula (Hill Notation) C12H13NO4

  • Molecular Weight 235.24

  •  Beilstein Registry Number 1581740

  •  MDL number MFCD00155428

  •  PubChem Substance ID 329818139

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Carbamates, Carbofuran metabolites, Insecticides,
grade   analytical standard
InChI Key   WXNZYYXXILQTKX-UHFFFAOYSA-N
form   neat
packaging   box of 1 ampule
mfr. no.   Chem Service, Inc., MET-754E
concentration   100 mg/ampule neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
FC2505000
Flash Point(F) 
>212 °F
Flash Point(C) 
>100 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?