• USA Home
  • ORGM-PB0.5 - Lead Organo Metallic Solution

EMAIL THIS PAGE TO A FRIEND
ORGM-PB0.5 Supelco

Lead Organo Metallic Solution

0.5000 wt. %

Synonym: Stabilized Organo-Metallic Oil Solutions

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Organic Pollutant Standards, Organometallic Compounds Standards, Petrochemical Standards,
grade   analytical standard
packaging   bottle of 100 g
concentration   0.5000 wt. %
application(s)   ICP/AAS: suitable
format   matrix material
storage temp.   room temp

Description

General description

Our organo-metallic oil standards are sulfur free, unless otherwise noted. The standards are stabilized with a proprietary chelation process and may be blended together to prepare multi-element solutions.-These standards are suitable for for X-ray Fluorescence (XRF), plasma emission (ICP or DCP), rotating dish (rotrode), or atomic absorption (AA).

Price and Availability


Nylon Membrane and Net Filters
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
>104 °F
Flash Point(C) 
>40 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?