EMAIL THIS PAGE TO A FRIEND
PBG7C Supelco

LEAD IN GASOLINE

analytical standard, set of 7 × 100 mL

Purchase

Properties

Related Categories ASTM D5059, ASTM Petroleum Standards, Analytical Standards, Analytical/Chromatography, Gasoline and Diesel,
grade   analytical standard
packaging   set of 7 × 100 mL
application(s)   ICP/AAS: suitable
suitability   suitable for D5059 per ASTM (Part C)
format   matrix material
storage temp.   room temp

Description

Analysis Note

Lead Standards in Various Matrices - Formulated sets
Standard No. Lead (Gms/Gal.)

Concentration Table: PBG7C
1 . . . . . . . . . 0.000
2 . . . . . . . . . 0.001
3 . . . . . . . . . 0.005
4 . . . . . . . . . 0.010
5 . . . . . . . . . 0.050
6 . . . . . . . . . 0.100
7 . . . . . . . . . 0.300

General description

7 standards per set; 100 mL each.

Price and Availability


Nylon Membrane and Net Filters
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1203 - class 3 - PG 2 - Gasoline
WGK Germany 
3
Flash Point(F) 
-49 °F
Flash Point(C) 
-45 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?