EMAIL THIS PAGE TO A FRIEND
PS1011 Supelco

Bentazon

analytical standard

  • CAS Number 25057-89-0

  • Empirical Formula (Hill Notation) C10H12N2O3S

  • Molecular Weight 240.28

  •  Beilstein Registry Number 530220

  •  EC Number 246-585-8

  •  MDL number MFCD00078640

  •  PubChem Substance ID 329823639

Purchase

Properties

grade   analytical standard
InChI Key   ZOMSMJKLGFBRBS-UHFFFAOYSA-N
form   neat
packaging   ampule of 250 mg
mfr. no.   Chem Service, Inc., PS-1011
concentration   250 mg neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
DK9900000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?