EMAIL THIS PAGE TO A FRIEND
PS1018 Supelco

Sulprofos

analytical standard

Synonym: Sulprophos

  • CAS Number 35400-43-2

  • Empirical Formula (Hill Notation) C12H19O2PS3

  • Molecular Weight 322.45

  •  Beilstein Registry Number 1990231

  •  EC Number 252-545-0

  •  MDL number MFCD00078721

  •  PubChem Substance ID 24899080

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Insecticides, Organophorous, Pesticides & Metabolites Standards More...
grade   analytical standard
InChI Key   JXHJNEJVUNHLKO-UHFFFAOYSA-N
form   neat
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., PS-1018
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2810 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis of 'Positive List' Chemicals (Mix 3) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of 'Positive List' Chemicals (Mix 3) on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Herbicides, Pesticides, Purification

Peer-Reviewed Papers
15

References

Related Products

replacement part

Product #

Description

Add to Cart

45665 Sulprofos, PESTANAL®, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?