• USA Home
  • PS101 - 2-Ethyl-2-butyl-1,3-propanediol

EMAIL THIS PAGE TO A FRIEND
PS101 Supelco

2-Ethyl-2-butyl-1,3-propanediol

analytical standard

Synonym: 2-Butyl-2-ethyl-1,3-propanediol

Purchase

Properties

Related Categories Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, E-G, Pesticides & Metabolites Standards More...
grade   analytical standard
InChI Key   DSKYSDCYIODJPC-UHFFFAOYSA-N
form   neat
packaging   ampule of 1000 mg
mfr. no.   Chem Service, Inc., PS-101
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   178 °C/50 mmHg(lit.)
mp   41-44 °C(lit.)
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
1
RTECS 
TY3605000
Flash Point(F) 
276.8 °F
Flash Point(C) 
136 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?