EMAIL THIS PAGE TO A FRIEND
PS1042 Supelco

Tetramethrin

analytical standard

  • CAS Number 7696-12-0

  • Empirical Formula (Hill Notation) C19H25NO4

  • Molecular Weight 331.41

  •  Beilstein Registry Number 8807938

  •  EC Number 231-711-6

  •  MDL number MFCD00084635

  •  PubChem Substance ID 24899093

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Insecticides, Pesticides & Metabolites Standards, Phthalimides,
grade   analytical standard
InChI Key   CXBMCYHAMVGWJQ-UHFFFAOYSA-N
form   neat
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., PS-1042
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS09  GHS09
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
GZ1730000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?