EMAIL THIS PAGE TO A FRIEND
PS1051 Supelco

Glyphosate

analytical standard

Synonym: N-(Phosphonomethyl)glycine, Glyphosate

  • CAS Number 1071-83-6

  • Linear Formula (HO)2P(O)CH2NHCH2CO2H

  • Molecular Weight 169.07

  •  Beilstein Registry Number 2045054

  •  EC Number 213-997-4

  •  MDL number MFCD00055350

Purchase

Properties

grade   analytical standard
InChI Key   XDDAORKBJWWYJS-UHFFFAOYSA-N
form   neat
packaging   ampule of 1000 mg
mfr. no.   Chem Service, Inc., PS-1051
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   230 °C (dec.)(lit.)
format   neat

Description

Testing Method

Purity determined by HPLC C18 method.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
MC1075000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?