EMAIL THIS PAGE TO A FRIEND
PS1064 Supelco

Baytoan

analytical standard

Synonym: α-tert-Butyl-β-(4-chlorophenoxy)-1H-1,2,4-triazole-1-ethanol, Triadimenol

  • CAS Number 55219-65-3

  • Empirical Formula (Hill Notation) C14H18ClN3O2

  • Molecular Weight 295.76

  •  Beilstein Registry Number 616470

  •  EC Number 259-537-6

  •  MDL number MFCD00055507

  •  PubChem Substance ID 24899105

Purchase

Properties

Related Categories A-B, All Fungicides, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
InChI Key   BAZVSMNPJJMILC-UHFFFAOYSA-N
form   neat
packaging   ampule of 250 mg
mfr. no.   Chem Service, Inc., PS-1064
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
KK2200000
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

HPLC Analysis of Triadimenol Enantiomers on Astec® CYCLOBOND® I 2000 HP-RSP

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Triadimenol Enantiomers on Astec® CYCLOBOND® I 2000 HP-RSP
Keywords: Chromatography, Environmental, Herbicides, High performance liquid chromatography, Pesticides, Pharmaceutical

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?