EMAIL THIS PAGE TO A FRIEND
PS1076 Supelco

Safrotin

analytical standard

Synonym: Propetamphos

  • CAS Number 31218-83-4

  • Empirical Formula (Hill Notation) C10H20NO4PS

  • Molecular Weight 281.31

  •  Beilstein Registry Number 1979853

  •  EC Number 250-517-2

  •  MDL number MFCD00137384

  •  PubChem Substance ID 24899112

Purchase

Properties

Related Categories Acaricides, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, Insecticides,
grade   analytical standard
InChI Key   BZNDWPRGXNILMS-VQHVLOKHSA-N
form   neat
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., PS-1076
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3278 6.1 / PGIII
WGK Germany 
3
RTECS 
GQ4750000

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis of Organophosphorus Pesticides (OP-Pest) in Cabbage, Onion, and Mushrooms on SLB-5ms after SPE using Supelclean™ ENVI-Carb™ II/PSA

From our library of Articles, Sigma-Aldrich presents GC Analysis of Organophosphorus Pesticides (OP-Pest) in Cabbage, Onion, and Mushrooms on SLB-5ms after SPE using Supelclean™ ENVI-Carb™ II/PSA
Keywords: Agriculture, Centrifugation, Chromatography, Degradations, Environmental, Gas chromatography, Nitrogen phosphorus detector, Pesticides, Purification, Sample preparations, Separation, Solid phase extractions

GC Analysis of a 19-Component Pesticide Mix on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents GC Analysis of a 19-Component Pesticide Mix on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Pesticides, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?