EMAIL THIS PAGE TO A FRIEND
PS1099 Supelco

Metalaxyl

analytical standard

Synonym: N-(2,6-Dimethylphenyl)-N-(methoxyacetyl)-DL-alanine methyl ester

  • CAS Number 57837-19-1

  • Empirical Formula (Hill Notation) C15H21NO4

  • Molecular Weight 279.33

  •  Beilstein Registry Number 2947777

  •  EC Number 260-979-7

  •  MDL number MFCD00055447

  •  PubChem Substance ID 24899122

Purchase

Properties

Related Categories Additional Standards, All Fungicides, Amide structure, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
InChI Key   ZQEIXNIJLIKNTD-UHFFFAOYSA-N
form   neat
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., PS-1099
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
AY6910000
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis of 'Positive List' Chemicals (Mix 4) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of 'Positive List' Chemicals (Mix 4) on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Herbicides, Pesticides, Purification

GC Analysis of Agricultural Pesticides on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of Agricultural Pesticides on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Herbicides, Metabolites, Pesticides, Purification, Size-exclusion chromatography

LC/MS Analysis of Pesticides on Ascentis® C18 (5 cm x 2.1 mm x 3 μm)

From our library of Articles, Sigma-Aldrich presents LC/MS Analysis of Pesticides on Ascentis® C18 (5 cm x 2.1 mm x 3 μm)
Keywords: Chromatography, Environmental, Environmental Analysis, Herbicides, High performance liquid chromatography, Mass spectrometry, Metabolites, Pesticides

SFC Analysis of Metalaxyl Enantiomers on Astec® Cellulose DMP

From our library of Articles, Sigma-Aldrich presents SFC Analysis of Metalaxyl Enantiomers on Astec® Cellulose DMP
Keywords: Agriculture, Chromatography, Fungicides, High performance liquid chromatography, Supercritical fluid chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?