• USA Home
  • PS1112 - Bentazon methyl derivative

EMAIL THIS PAGE TO A FRIEND
PS1112 Supelco

Bentazon methyl derivative

analytical standard

Synonym: N-Methylbentazon

  • CAS Number 61592-45-8

  • Empirical Formula (Hill Notation) C11H14N2O3S

  • Molecular Weight 254.31

  •  MDL number MFCD00152260

  •  PubChem Substance ID 24899126

Purchase

Properties

Related Categories A-D, All Herbicide Standards, Analytical Standards, Analytical/Chromatography, Bentazon metabolite,
grade   analytical standard
InChI Key   XFTQFXBQDVWOCY-UHFFFAOYSA-N
form   neat
packaging   ampule of 50 mg
mfr. no.   Chem Service, Inc., PS-1112
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
3
RTECS 
DK9900000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?