• USA Home
  • PS1202 - Azamethiphos solution

EMAIL THIS PAGE TO A FRIEND
PS1202 Supelco

Azamethiphos solution

100 μg/mL in methylene chloride, analytical standard

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Insecticides, Organophorous, Pesticides & Metabolites Standards More...
grade   analytical standard
packaging   ampule of 5 mL
mfr. no.   Chem Service, Inc., PS-1202
concentration   100 μg/mL in methylene chloride
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Target organs 
Blood, Central nervous system, Liver
RIDADR 
UN 1593 6.1 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?