EMAIL THIS PAGE TO A FRIEND
PS2012 Supelco

Thiodicarb

analytical standard

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Carbamates, Insecticides, Molluscicides,
grade   analytical standard
InChI Key   XDOTVMNBCQVZKG-OXAWKVHCSA-N
form   neat
packaging   ampule of 250 mg
mfr. no.   Chem Service, Inc., PS-2012
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN2811 - class 6.1 - PG 2 - RQ - Toxic solids, organic, n.o.s. HI: all
WGK Germany 
3
RTECS 
KJ4301050

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis of Agricultural Pesticides on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of Agricultural Pesticides on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Herbicides, Metabolites, Pesticides, Purification, Size-exclusion chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?